This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALKBH7 SIRNA Assays ›
Gene Symbol:
Alkbh7
Gene Name:
alkB, alkylation repair homolog 7 (E. coli)
Gene Aliases:
2310045B01Rik, 2510008E23Rik, Abh7, Spata11
Chromosome Location:
Chr.17: 56997339 - 56999340 on Build GRCm38
UniGene ID:
Mm.196150
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s83180

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alkbh7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025538.3 NP_079814.1 1 173
NM_027372.1 NP_081648.1 1 173
GenBank mRNA AK007380.1 - 1 151
AK009812.1 - 1 160
AK010930.1 - 1 174
BC029677.1 - 1 140 5364485

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
cellular response to DNA damage stimulus
regulation of lipid storage
programmed cell death
oxidation-reduction process
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
sulfonate dioxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header