This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LBHD1 SIRNA Assays ›
Gene Symbol:
Lbhd1 1810009A15Rik
Gene Name:
LBH domain containing 1
RIKEN cDNA 1810009A15 gene
Gene Aliases:
Gm21743
-
Chromosome Location:
Chr.19: 8883878 - 8890759 on Build GRCm38
Chromosome Location:
Chr.19: 8888883 - 8890759 on Build GRCm38
UniGene ID:
Mm.27503
Mm.27503
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s82942

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lbhd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286103.1 NP_001273032.1 4 615
GenBank mRNA CK619869.1 - 4 585
Gene Symbol: 1810009A15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025463.3 NP_079739.1 2 119
GenBank mRNA AK007390.1 - 2 100
AK012061.1 - 2 109
BC047099.1 - 2 98 5253822
BC058702.1 - 2 82 6822952
BY264380.1 - 2 121
BY273399.1 - 1 262

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0