This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GHITM SIRNA Assays ›
Gene Symbol:
Ghitm
Gene Name:
growth hormone inducible transmembrane protein
Gene Aliases:
1010001P14Rik, C77840, MICS1, PTD010, Tmbim5
Chromosome Location:
Chr.14: 37120444 - 37135327 on Build GRCm38
UniGene ID:
Mm.182912
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s82564

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ghitm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199122.1 NP_001186051.1 8 1059
NM_078478.5 NP_510963.1 8 1012
GenBank mRNA AF412297.1 - 8 956
AK005636.1 - 8 957
AK093887.1 - 8 988
AK134592.1 - 8 942
AK146433.1 - 8 919
AK146853.1 - 8 975
AK151488.1 - 8 989
AK158904.1 - 8 1002
AK159695.1 - 8 986
AK159724.1 - 8 986
AK164122.1 - 8 957
AK167970.1 - 8 983
AK168022.1 - 8 989
AK168678.1 - 8 1060
AK168800.1 - 8 968
AK189963.1 - 2 177
BC008622.1 - 8 990 3588758
BC010224.1 - 8 992 3500685

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001005908, XM_006252786, FQ217853, BC083778, FQ212260.

Gene Ontology Categories:

Function(s)

apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0