This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SP5 SIRNA Assays ›
Gene Symbol:
Sp5
Gene Name:
trans-acting transcription factor 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 70474923 - 70477729 on Build GRCm38
UniGene ID:
Mm.423397
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s82226

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sp5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022435.2 NP_071880.1 2 1082
GenBank mRNA AF279479.1 - 2 839
AK036234.1 - 2 1082
BC116691.1 - 2 933 40106236
BC117505.1 - 2 933 40106237

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106552.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
post-anal tail morphogenesis
bone morphogenesis
cellular response to organic cyclic compound
RNA polymerase II regulatory region sequence-specific DNA binding
nucleic acid binding
DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0