This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SUMF1 SIRNA Assays ›
Gene Symbol:
Sumf1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AA543204, AI463102, AI851573, FGE
Chromosome Location:
Chr.6: 108107021 - 108185591 on Build GRCm38
UniGene ID:
Mm.439876
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s81725

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sumf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145937.3 NP_666049.2 4 561
GenBank mRNA AK151874.1 - 4 562
AK159192.1 - 4 550
AK160917.1 - 4 543
AK161203.1 - 4 554
AK194677.1 - 3 140
AK195132.1 - 3 140
AK198586.1 - 3 140
BC026981.1 - 4 534 5365580

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
protein binding
dimethyl sulfoxide reductase activity
oxidoreductase activity
tetrachloro-p-hydroquinone reductive dehalogenase activity
protein homodimerization activity
metal ion binding
trichloro-p-hydroquinone reductive dehalogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0