This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR1507 SIRNA Assays ›
Gene Symbol:
Olfr1507
Gene Name:
olfactory receptor 1507
Gene Aliases:
MOR244-1, Mor28, Or28
Chromosome Location:
Chr.14: 52489935 - 52495695 on Build GRCm38
UniGene ID:
Mm.88822
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s81172

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1507
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170918.1 NP_001164389.1 2 1059
NM_020512.2 NP_065258.2 1 566
GenBank mRNA BC119163.1 - 1 566 8733912
BC119165.1 - 1 566 8733914

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000102.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header