This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FADS2 SIRNA Assays ›
Gene Symbol:
Fads2
Gene Name:
fatty acid desaturase 2
Gene Aliases:
2900042M13Rik, Fadsd2
Chromosome Location:
Chr.19: 10064164 - 10101503 on Build GRCm38
UniGene ID:
Mm.38901
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s80652

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fads2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019699.1 NP_062673.1 4 660
GenBank mRNA AF126798.1 - 4 660
AK031337.1 - 4 773
AK083282.1 - 627
AK143524.1 - 678
BC057189.1 - 4 648 5137343
HQ264057.1 - 4 553
HQ264058.1 - 4 553

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
transport
oxidation-reduction process
stearoyl-CoA 9-desaturase activity
acyl-CoA desaturase activity
oxidoreductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0