This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BACE2 SIRNA Assays ›
Gene Symbol:
Bace2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
1110059C24Rik, AEPLC, AI850424, ALP56, ARP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.16: 97356728 - 97439012 on Build GRCm38
UniGene ID:
Mm.97885
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s80111

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bace2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019517.4 NP_062390.3 6, 7 1044
GenBank mRNA AF051150.1 - 2, 3 193
AF216310.1 - 6, 7 995
AK052309.1 - 6, 7 1031
AK078770.1 - 6, 7 1011
AK165036.1 - 6, 7 980
BC120773.1 - 6, 7 1010 40129696
BC131947.1 - 6, 7 1010 40130224

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity
peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0