This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CTS8 SIRNA Assays ›
Gene Symbol:
Cts8 Cts8-ps
Gene Name:
cathepsin 8
cathepsin 8, pseudogene
Gene Aliases:
CTS2, Epcs68, Epcs70
Cts2-ps
Chromosome Location:
Chr.13: 61246747 - 61255348 on Build GRCm38
Chromosome Location:
Chr.13: 61281380 - 61288850 on Build GRCm38
UniGene ID:
Mm.342861
Mm.475565
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s80091

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cts8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019541.3 NP_062414.3 5 687
GenBank mRNA AF250840.1 - 5 699
AK049474.1 - 5 688
AY014780.1 - 5 686
BC068241.1 - 5 677 30019120
Gene Symbol: Cts8-ps
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC050022.1 - 3 2376

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood vessel remodeling
proteolysis
protein catabolic process
proteolysis involved in cellular protein catabolic process
trophoblast giant cell differentiation
cysteine-type endopeptidase activity
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0