This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF3D SIRNA Assays ›
Gene Symbol:
Eif3d
Gene Name:
eukaryotic translation initiation factor 3, subunit D
Gene Aliases:
66/67kDa, AA407891, Eif3s7, eIF3p66
Chromosome Location:
Chr.15: 77958998 - 77971088 on Build GRCm38
UniGene ID:
Mm.3955
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s79901

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif3d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018749.2 NP_061219.2 9 978
GenBank mRNA AB012580.1 - 9 919
AK045228.1 - 9 920
AK049767.1 - 9 917
AK145689.1 - 9 933
BC018598.1 - 9 916
BC089020.1 - 9 912 30551991

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

formation of cytoplasmic translation initiation complex
translation
translational initiation
translation initiation factor activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0