This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATF7IP SIRNA Assays ›
Gene Symbol:
Atf7ip
Gene Name:
activating transcription factor 7 interacting protein
Gene Aliases:
2610204M12Rik, AM, Mcaf1
Chromosome Location:
Chr.6: 136518789 - 136610850 on Build GRCm38
UniGene ID:
Mm.173271
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s79557

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atf7ip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019426.2 NP_062299.2 2 1286
GenBank mRNA AJ132702.1 - 2 1279
AK134252.1 - 2 1293
AK167542.1 - 3 1332
BC022714.1 - 2 1279
BC037617.1 - 1288
BC048827.1 - 3 1447
BC052460.1 - 2 1286 6834550
BC060246.1 - 2 1269 5705945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
regulation of RNA polymerase II transcriptional preinitiation complex assembly
transcription corepressor activity
ATPase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0