This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TXNL1 SIRNA Assays ›
Gene Symbol:
Txnl1
Gene Name:
thioredoxin-like 1
Gene Aliases:
32kDa, TRP32, Txnl
Chromosome Location:
Chr.18: 63662801 - 63692359 on Build GRCm38
UniGene ID:
Mm.19169
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s79160

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Txnl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016792.4 NP_058072.2 3 529
GenBank mRNA AF052660.1 - 3 358
AK029807.1 - 3 530
AK051698.1 - 3 504
AK144211.1 - 3 486
AK150326.1 - 3 492
AK150887.1 - 3 498
AK150983.1 - 3 498
AK152101.1 - 3 498
AK167954.1 - 3 500
AK168712.1 - 3 493
AK209072.1 - 1 95
AK212711.1 - 1 111
BC061123.1 - 3 556 6596299

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sulfate assimilation
protein folding
glycerol ether metabolic process
transport
cellular response to oxidative stress
cell redox homeostasis
oxidation-reduction process
protein disulfide oxidoreductase activity
disulfide oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0