This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLIC4 SIRNA Assays ›
Gene Symbol:
Clic4
Gene Name:
chloride intracellular channel 4 (mitochondrial)
Gene Aliases:
D0Jmb3, TU-74, mc3s5, mtCLIC
Chromosome Location:
Chr.4: 135213969 - 135272760 on Build GRCm38
UniGene ID:
Mm.257765
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s78124

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clic4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013885.2 NP_038913.1 6 796
GenBank mRNA AF102578.1 - 6 794
AK031211.1 - 6 803
AK049191.1 - 6 801
AK049255.1 - 6 802
AK150993.1 - 6 800
AK151504.1 - 2 72
AK152209.1 - 2 72
AK152273.1 - 6 802
AK153432.1 - 6 803
AK155826.1 - 6 800
AK159402.1 - 6 803
AK160856.1 - 6 802
AK161034.1 - 6 801
AK166244.1 - 6 804
AK166794.1 - 6 800
AK169376.1 - 6 800
AK169834.1 - 6 800
AK170942.1 - 6 851
AK171243.1 - 6 803
AK172026.1 - 6 802
AK172516.1 - 6 776
AK172660.1 - 6 800
BC046384.1 - 6 780 3155573
BC052890.1 - 6 784 30056606
CT010279.1 - 6 625

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
endothelial cell morphogenesis
transport
ion transport
chloride transport
apoptotic process
vacuolar acidification
fertilization
keratinocyte differentiation
negative regulation of cell migration
regulation of ion transmembrane transport
multicellular organism growth
branching morphogenesis of an epithelial tube
retina vasculature morphogenesis in camera-type eye
cellular response to calcium ion
chloride transmembrane transport
voltage-gated ion channel activity
chloride channel activity
protein binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0