This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SGCB SIRNA Assays ›
Gene Symbol:
Sgcb
Gene Name:
sarcoglycan, beta (dystrophin-associated glycoprotein)
Gene Aliases:
43kDa, AI747103, AI844814, beta-SG
Chromosome Location:
Chr.5: 73632749 - 73647731 on Build GRCm38
UniGene ID:
Mm.89310
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76827

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sgcb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011890.4 NP_036020.1 2 118
GenBank mRNA AK014381.2 - 2 132
AK132306.1 - 2 177
AK169403.1 - 2 119
BC052349.1 - 2 108 6744003

Back To Top

More Information


Gene Ontology Categories:

Function(s)

muscle organ development
muscle fiber development
cardiac muscle cell development
membrane organization
vascular smooth muscle cell development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0