This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF13 SIRNA Assays ›
Gene Symbol:
Rnf13
Gene Name:
ring finger protein 13
Gene Aliases:
2010001H16Rik, Rzf
Chromosome Location:
Chr.3: 57736043 - 57835233 on Build GRCm38
UniGene ID:
Mm.274360
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76803

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF037205.1 - 6 533
AF037206.1 - 6 533
AK034135.1 - 6 594
AK036591.1 - 6 578
AK048381.1 - 1 1679
AK050095.1 - 6 1278
AK082296.1 - 6 574
AK139447.1 - 6 539
AK140682.1 - 6 588
AK158046.1 - 6 574
AK167278.1 - 6 538
BC058182.1 - 6 541 5324040

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0