This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMD13 SIRNA Assays ›
Gene Symbol:
Psmd13
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 13
Gene Aliases:
S11
Chromosome Location:
Chr.7: 140882394 - 140898642 on Build GRCm38
UniGene ID:
Mm.29760
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76774

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011875.4 NP_036005.1 7 487
GenBank mRNA AF107838.1 - 7 448
AK051078.1 - 7 478
AK075936.1 - 7 488
AK081160.1 - 7 485
AK145654.1 - 7 450
AK163017.1 - 7 482
AK167169.1 - 7 472
AK171504.1 - 7 468
AK206994.1 - 7 363
BC029989.1 - 7 427
BC139184.1 - 7 465 8862206
BC139187.1 - 7 465 8862209

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108925, BC158732.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
meiosis I
endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0