This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WBP4 SIRNA Assays ›
Gene Symbol:
Wbp4
Gene Name:
WW domain binding protein 4
Gene Aliases:
AW545037, BB101031, FBP21
Chromosome Location:
Chr.14: 79459937 - 79481268 on Build GRCm38
UniGene ID:
Mm.476813
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76027

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wbp4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018765.3 NP_061235.2 7 854
GenBank mRNA AF071184.1 - 7 706
AK135215.1 - 7 854
AK159522.1 - 7 1106
AK160559.1 - 7 591
AK161496.1 - 7 548
AK162009.1 - 7 596
AK164984.1 - 1 2087
BC034851.1 - 7 549 1399984
U40746.1 - 3 134

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
mRNA cis splicing, via spliceosome
nucleic acid binding
protein binding
zinc ion binding
metal ion binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header