This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPV2 SIRNA Assays ›
Gene Symbol:
Trpv2
Gene Name:
transient receptor potential cation channel, subfamily V, member 2
Gene Aliases:
GRC, OTRPC2, VRL-1, Vrl1
Chromosome Location:
Chr.11: 62574486 - 62600305 on Build GRCm38
UniGene ID:
Mm.288064
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s76000

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trpv2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011706.2 NP_035836.2 11 1895
GenBank mRNA AB021665.1 - 11 1977
AK008689.1 - 9 1420
AK089004.1 - 12 1910
BC005415.1 - 10 1842 3487527

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
calcium ion transport
apoptotic process
negative regulation of cell proliferation
response to temperature stimulus
response to heat
positive regulation of axon extension
transmembrane transport
calcium ion transmembrane transport
positive regulation of calcium ion import
cation transmembrane transport
ion channel activity
cation channel activity
calcium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0