This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2H SIRNA Assays ›
Gene Symbol:
Ube2h
Gene Name:
ubiquitin-conjugating enzyme E2H
Gene Aliases:
1500009C23Rik, AI181839, AI326965, AI462483, AW227540, E2-20K, Gid3, N28148, Ubc8
Chromosome Location:
Chr.6: 30211289 - 30304539 on Build GRCm38
UniGene ID:
Mm.332967
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75691

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ube2h
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169576.1 NP_001163047.1 2 539
NM_009459.3 NP_033485.1 2 539
GenBank mRNA AK131688.1 - 2 517
AK161414.1 - 2 523
BC008517.1 - 2 345 3600233
U19854.1 - 2 268

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
protein K11-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0