This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATP6V0A2 SIRNA Assays ›
Gene Symbol:
Atp6v0a2
Gene Name:
ATPase, H+ transporting, lysosomal V0 subunit A2
Gene Aliases:
8430408C20Rik, AI385560, ATP6a2, AW489264, Atp6n1d, Atp6n2, C76904, ISF, J6B7, SHIF, Stv1, TJ6M, TJ6s, Tj6, V-ATPase 116 kDa, V-ATPase a2
Chromosome Location:
Chr.5: 124629052 - 124646816 on Build GRCm38
UniGene ID:
Mm.1158
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75170

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp6v0a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011596.5 NP_035726.2 925
GenBank mRNA AB022323.1 - 891
AF218252.1 - 910
AK155055.1 - 902
AK170122.1 - 1179
BC018369.1 - 878
BC108991.1 - 822 40047832
BC108992.1 - 822 40047833
BC112905.1 - 885 3670722
M31226.1 - 821
X55184.1 - 822

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053775, DQ286425, BC168659.

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
vacuolar acidification
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
vacuolar proton-transporting V-type ATPase complex assembly
protein binding
hydrogen ion transmembrane transporter activity
proton-transporting ATPase activity, rotational mechanism
ATPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0