This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THRB SIRNA Assays ›
Gene Symbol:
Thrb
Gene Name:
thyroid hormone receptor beta
Gene Aliases:
Nr1a2, T3R[b], T3Rbeta, Thrb1, Thrb2, c-erbAbeta
Chromosome Location:
Chr.14: 17659218 - 18040097 on Build GRCm38
UniGene ID:
Mm.32563
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75116

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thrb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113417.1 NP_001106888.1 8 1000
NM_009380.3 NP_033406.1 5 1086
GenBank mRNA AB221613.1 - 5 853
AK133662.1 - 5 583
AK158826.1 - 8 1001
BC089035.1 - 5 1086 30695885
BC119552.1 - 5 1023 40057420
BC119553.1 - 5 1027 40057421
S62756.1 - 7 870

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
positive regulation of cell proliferation
organ morphogenesis
regulation of lipid metabolic process
intracellular receptor signaling pathway
positive regulation of chondrocyte differentiation
steroid hormone mediated signaling pathway
positive regulation of ossification
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
Type I pneumocyte differentiation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
enzyme binding
chromatin DNA binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
thyroid hormone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0