This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP2R5D SIRNA Assays ›
Gene Symbol:
Ppp2r5d
Gene Name:
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases:
B'delta, TEG-271, Tex271
Chromosome Location:
Chr.17: 46682991 - 46705020 on Build GRCm38
UniGene ID:
Mm.295009
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75009

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp2r5d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009358.3 NP_033384.2 7 859
GenBank mRNA AB053464.1 - 7 804
AF316616.2 - 7 716
AK160587.1 - 7 800
AY338234.1 - 7 789
BC010716.1 - 7 772 2655861

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
positive regulation of neuron projection development
regulation of protein autophosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of neurotrophin TRK receptor signaling pathway
cellular response to growth factor stimulus
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header