This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TEX264 SIRNA Assays ›
Gene Symbol:
Tex264
Gene Name:
testis expressed gene 264
Gene Aliases:
TEG-264
Chromosome Location:
Chr.9: 106658746 - 106685952 on Build GRCm38
UniGene ID:
Mm.22266
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s75003

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tex264
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081654.2 NP_001075123.1 4 564
NM_001286498.1 NP_001273427.1 4 764
NM_011573.3 NP_035703.2 4 1019
GenBank mRNA AK003113.1 - 4 527
AK009326.1 - 4 870
AK159046.1 - 3 1384
AK219517.1 - 2 297
BC002248.1 - 4 528 3493141
BC006608.1 - 4 502 3157453
CX220909.1 - 4 532
DV656490.1 - 3 217
DV661702.1 - 2 339

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header