This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TERT SIRNA Assays ›
Gene Symbol:
Tert
Gene Name:
telomerase reverse transcriptase
Gene Aliases:
EST2, TCS1, TP2, TR, TRT
Chromosome Location:
Chr.13: 73626797 - 73649843 on Build GRCm38
UniGene ID:
Mm.10109
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74977

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tert
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009354.1 NP_033380.1 8 2463
GenBank mRNA AF051911.1 - 8 2463
AF073311.1 - 8 2434
BC082327.1 - 9 1965
BC127068.1 - 9 1751 40110173
FJ210936.1 - 4 459

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, RNA-templated
RNA-dependent DNA biosynthetic process
telomere maintenance via telomerase
mitochondrion organization
negative regulation of gene expression
DNA strand elongation
positive regulation of Wnt signaling pathway
production of siRNA involved in RNA interference
RNA biosynthetic process
positive regulation of hair cycle
positive regulation of nitric-oxide synthase activity
cellular response to hypoxia
DNA biosynthetic process
replicative senescence
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of production of siRNA involved in RNA interference
positive regulation of stem cell proliferation
negative regulation of cellular senescence
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
tRNA binding
transcription coactivator binding
DNA binding
telomerase activity
telomerase RNA reverse transcriptase activity
RNA binding
RNA-directed DNA polymerase activity
RNA-directed RNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
telomeric DNA binding
protein homodimerization activity
metal ion binding
telomerase RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header