This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBX5 SIRNA Assays ›
Gene Symbol:
Tbx5
Gene Name:
T-box 5
Gene Aliases:
-
Chromosome Location:
Chr.5: 119796515 - 119885220 on Build GRCm38
UniGene ID:
Mm.103636
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74783

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbx5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011537.3 NP_035667.1 4 939
GenBank mRNA AF140427.1 - 4 704
BC090639.1 - 4 943 30548234
U57330.1 - 2 100

Back To Top

More Information


Panther Classification:

Molecular Function -

Rel homology transcription factor

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
bundle of His development
atrioventricular valve morphogenesis
endocardial cushion development
cardiac left ventricle formation
ventricular cardiac muscle tissue development
ventricular septum development
atrial septum development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cell-cell signaling
multicellular organism development
pattern specification process
heart development
negative regulation of cell proliferation
lung development
embryonic limb morphogenesis
negative regulation of cell migration
embryonic forelimb morphogenesis
forelimb morphogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cardioblast differentiation
cardiac muscle cell differentiation
pericardium development
negative regulation of cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
transdifferentiation
atrial septum morphogenesis
positive regulation of secondary heart field cardioblast proliferation
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0