This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYNJ2 SIRNA Assays ›
Gene Symbol:
Synj2
Gene Name:
synaptojanin 2
Gene Aliases:
AI481647, SJ2, mKIAA0348
Chromosome Location:
Chr.17: 5941280 - 6045553 on Build GRCm38
UniGene ID:
Mm.236068
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74661

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Synj2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113351.1 NP_001106822.1 11 1711
NM_001113352.2 NP_001106823.1 12 1800
NM_001113353.2 NP_001106824.1 12 1800
NM_001290698.1 NP_001277627.1 6 828
NM_011523.2 NP_035653.2 11 1711
GenBank mRNA AF041857.1 - 12 1801
AF041858.1 - 1 63
AF041859.1 - 5 592
AF041862.1 - 11 1656
AK014584.1 - 8 955
AK019694.1 - 6 829
AK029755.1 - 9 1218
AK038038.1 - 9 1124
AK129122.1 - 10 1228
AK142919.1 - 11 1432
AK148053.1 - 12 1538
AK161467.1 - 6 812
AK163512.1 - 10 1441
BC058749.1 - 12 1677 6390688
BC060214.1 - 11 1557 6838657

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
intracellular distribution of mitochondria
nucleotide binding
nucleic acid binding
RNA binding
phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
inositol-polyphosphate 5-phosphatase activity
hydrolase activity
SH3 domain binding
PDZ domain binding
phosphoric ester hydrolase activity
phosphatidylinositol monophosphate phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0