This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC19A1 SIRNA Assays ›
Gene Symbol:
Slc19a1
Gene Name:
solute carrier family 19 (folate transporter), member 1
Gene Aliases:
AI323572, RFC, RFC-1, RFC1
Chromosome Location:
Chr.10: 77032269 - 77050432 on Build GRCm38
UniGene ID:
Mm.265060
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s73871

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc19a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199271.1 NP_001186200.1 5 1239
NM_031196.3 NP_112473.1 5 1333
GenBank mRNA AK002560.1 - 5 1250
AK050714.1 - 5 1363
AK079592.1 - 5 1294
AK085024.2 - 5 1281
AK088803.1 - 5 1296
AK138577.1 - 5 1159
BC015263.1 - 5 1251 4239690
L23755.1 - 4 1191
L36539.1 - 5 1246
U32469.1 - 5 1273
U66103.1 - 5 1286

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
folic acid transport
methotrexate transport
transmembrane transport
folic acid binding
folic acid transporter activity
reduced folate carrier activity
methotrexate transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0