This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RFX3 SIRNA Assays ›
Gene Symbol:
Rfx3
Gene Name:
regulatory factor X, 3 (influences HLA class II expression)
Gene Aliases:
C230093O12Rik, MRFX3
Chromosome Location:
Chr.19: 27761718 - 28011698 on Build GRCm38
UniGene ID:
Mm.313321
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s72918

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rfx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166414.1 NP_001159886.1 4 358
NM_011265.3 NP_035395.2 3 399
GenBank mRNA AK030008.1 - 5 431
AK041120.1 - 4 459
AK140928.1 - 3 282
BC017598.1 - 3 373 4483833

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012172, XM_006231235, XM_008760340, XM_006231233, XM_008760341, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
determination of left/right symmetry
cell differentiation
endocrine pancreas development
cilium assembly
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
regulation of insulin secretion
cilium-dependent cell motility
epithelial cilium movement involved in determination of left/right asymmetry
type B pancreatic cell maturation
positive regulation of type B pancreatic cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0