This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBM6 SIRNA Assays ›
Gene Symbol:
Rbm6
Gene Name:
RNA binding motif protein 6
Gene Aliases:
4930506F14Rik, Def-3, NY-LU-12, g16, mKIAA4015
Chromosome Location:
Chr.9: 107773559 - 107872819 on Build GRCm38
UniGene ID:
Mm.139926
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s72783

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rbm6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011251.3 NP_035381.2 3 634
NM_029169.3 NP_083445.1 3 905
GenBank mRNA AJ006486.2 - 3 513
AK077899.1 - 3 811
AK081846.1 - 3 559
AK134164.1 - 3 545
AK145612.1 - 3 794
AK220446.1 - 3 783
BC026129.1 - 3 659 3982861
BC026130.1 - 3 808

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108186, XM_006243781, XM_008766534, XM_006243780.

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0