This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTEN SIRNA Assays ›
Gene Symbol:
Pten
Gene Name:
phosphatase and tensin homolog
Gene Aliases:
2310035O07Rik, A130070J02Rik, AI463227, B430203M17Rik, MMAC1, TEP1
Chromosome Location:
Chr.19: 32757577 - 32826160 on Build GRCm38
UniGene ID:
Mm.245395
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s72351

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pten
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008960.2 NP_032986.1 9 2021
GenBank mRNA AK030750.1 - 5 784
AK076980.1 - 9 1979
AK088717.1 - 9 2000
AK138025.1 - 2 330
AK148736.1 - 9 1856
BC021445.1 - 9 1370 5038272
U92437.1 - 9 2101

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_000314, CR450306, JF268690, U92436, BC038293, and others.
This siRNA also targets the following transcripts in Rat: NM_031606, AF017185, AF455569.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of protein phosphorylation
regulation of B cell apoptotic process
protein dephosphorylation
lipid metabolic process
apoptotic process
neuron-neuron synaptic transmission
nervous system development
synapse assembly
central nervous system development
heart development
learning or memory
memory
locomotory behavior
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of neuron projection development
dephosphorylation
cell migration
dentate gyrus development
central nervous system neuron axonogenesis
negative regulation of cell migration
neuron projection development
negative regulation of myelination
regulation of protein stability
negative regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
regulation of synaptic transmission, GABAergic
central nervous system myelin maintenance
regulation of cellular component size
regulation of myeloid cell apoptotic process
multicellular organismal response to stress
social behavior
peptidyl-tyrosine dephosphorylation
cellular response to decreased oxygen levels
maternal behavior
positive regulation of apoptotic process
negative regulation of apoptotic process
protein kinase B signaling
endothelial cell migration
locomotor rhythm
negative regulation of cell size
negative regulation of organ growth
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
platelet-derived growth factor receptor signaling pathway
regulation of axon regeneration
cardiac muscle tissue development
forebrain morphogenesis
brain morphogenesis
negative regulation of epithelial cell proliferation
negative regulation of phagocytosis
negative regulation of axonogenesis
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of cell cycle
negative regulation of focal adhesion assembly
negative regulation of protein kinase B signaling
rhythmic synaptic transmission
canonical Wnt signaling pathway
synapse maturation
prepulse inhibition
male mating behavior
long-term synaptic potentiation
long term synaptic depression
prostate gland growth
dendritic spine morphogenesis
negative regulation of dendritic spine morphogenesis
positive regulation of ERK1 and ERK2 cascade
cellular response to hypoxia
negative regulation of ribosome biogenesis
negative regulation of cell aging
negative regulation of excitatory postsynaptic potential
presynaptic membrane assembly
postsynaptic density assembly
positive regulation of ubiquitin protein ligase activity
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of G1/S transition of mitotic cell cycle
positive regulation of excitatory postsynaptic potential
negative regulation of synaptic vesicle clustering
positive regulation of apoptotic signaling pathway
magnesium ion binding
phosphatidylinositol-3-phosphatase activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
platelet-derived growth factor receptor binding
protein binding
protein tyrosine/serine/threonine phosphatase activity
anaphase-promoting complex binding
phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
hydrolase activity
phosphatase activity
enzyme binding
protein kinase binding
PDZ domain binding
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
phosphatidylinositol-3,4-bisphosphate 3-phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0