This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMB10 SIRNA Assays ›
Gene Symbol:
Psmb10
Gene Name:
proteasome (prosome, macropain) subunit, beta type 10
Gene Aliases:
Mecl-1, Mecl1
Chromosome Location:
Chr.8: 105935728 - 105938454 on Build GRCm38
UniGene ID:
Mm.787
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s72277

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmb10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013640.3 NP_038668.2 3 589
GenBank mRNA AK144096.1 - 3 312
BC004730.1 - 3 315 3583052
CT010274.1 - 3 207
D85561.1 - 3 284
U77784.1 - 3 276
Y10875.1 - 3 307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
proteolysis
proteasomal ubiquitin-independent protein catabolic process
T cell proliferation
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0