This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLN SIRNA Assays ›
Gene Symbol:
Pln
Gene Name:
phospholamban
Gene Aliases:
Plb
Chromosome Location:
Chr.10: 53337686 - 53345999 on Build GRCm38
UniGene ID:
Mm.34145
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s71847

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pln
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141927.1 NP_001135399.1 3 960
NM_023129.5 NP_075618.1 2 833
GenBank mRNA AF214653.1 - 1 74
AK002622.1 - 2 813
AK040718.1 - 2 804
AK052199.1 - 2 767
AK142708.1 - 2 788
S46792.1 - 2 807

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
calcium ion transport
cellular calcium ion homeostasis
Notch signaling pathway
regulation of heart contraction
response to zinc ion
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
response to testosterone
negative regulation of catalytic activity
negative regulation of heart contraction
cardiac muscle tissue development
protein homooligomerization
regulation of ryanodine-sensitive calcium-release channel activity
regulation of cardiac muscle cell contraction
adrenergic receptor signaling pathway involved in heart process
regulation of the force of heart contraction by cardiac conduction
regulation of calcium ion import
regulation of relaxation of muscle
calcium channel regulator activity
protein binding
ATPase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0