siRNA Details
Gene Symbol: Plcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021280.3 | NP_067255.2 | 11, 12 | 1137 | ||
GenBank mRNA | AK134041.1 | - | 10, 11 | 942 | ||
BC057161.1 | - | 11, 12 | 1147 | |||
BC065091.1 | - | 11, 12 | 1137 | 6854923 |
