This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PFKM SIRNA Assays ›
Gene Symbol:
Pfkm
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
AI131669, ATP-PFK, PFK-A, PFK-M, Pfk-4, Pfk4, Pfka, Pfkx
Chromosome Location:
Chr.15: 98092589 - 98132447 on Build GRCm38
UniGene ID:
Mm.272582
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s71521

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pfkm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163487.1 NP_001156959.1 13 1315
NM_001163488.1 NP_001156960.1 13 1251
NM_021514.4 NP_067489.3 12 1268
GenBank mRNA AF249894.1 - 12 1208
AK002711.2 - 13 1218
AK049773.1 - 13 1223
BC005526.1 - 12 1232 3495173
D21866.1 - 2 79

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001166688, NM_001166686, NM_000289, NM_001166687, CR541698, and others.
This siRNA also targets the following transcripts in Rat: NM_031715, BC094212, FQ211719, U25651, FN433010, and others.

Gene Ontology Categories:

Function(s) Process(es)

fructose 6-phosphate metabolic process
glycolytic process
metabolic process
phosphorylation
fructose 1,6-bisphosphate metabolic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
protein homotetramerization
glycolytic process through fructose-6-phosphate
nucleotide binding
catalytic activity
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
phosphofructokinase activity
AMP binding
kinase activity
transferase activity
kinase binding
carbohydrate binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
fructose-6-phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0