This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BLOC1S6 SIRNA Assays ›
Gene Symbol:
Bloc1s6
Gene Name:
biogenesis of lysosomal organelles complex-1, subunit 6, pallidin
Gene Aliases:
BLOC-1, Pldn, Stx13bp1, pa
Chromosome Location:
Chr.2: 122738505 - 122749487 on Build GRCm38
UniGene ID:
Mm.268921
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s71209

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bloc1s6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019788.3 NP_062762.1 3 371
GenBank mRNA AK038381.1 - 3 349
AK077720.1 - 3 353
AK147116.1 - 3 374
AK160580.1 - 3 341
AK170964.1 - 3 302
AK172571.1 - 3 352
BC016554.1 - 3 350
BC085260.1 - 3 340 6437935

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025714, BC097469.

Gene Ontology Categories:

Function(s) Process(es)

vesicle docking involved in exocytosis
vesicle fusion
blood coagulation
anterograde axonal transport
melanocyte differentiation
neuron projection development
melanosome transport
positive regulation of natural killer cell activation
secretion of lysosomal enzymes
endosome to melanosome transport
pigmentation
anterograde synaptic vesicle transport
positive regulation of pigment cell differentiation
membrane fusion
protein binding
identical protein binding
protein homodimerization activity
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header