This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other P2RY1 SIRNA Assays ›
Gene Symbol:
P2ry1
Gene Name:
purinergic receptor P2Y, G-protein coupled 1
Gene Aliases:
P2Y1
Chromosome Location:
Chr.3: 61002785 - 61008982 on Build GRCm38
UniGene ID:
Mm.281452
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s71195

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: P2ry1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282016.1 NP_001268945.1 1 1378
NM_008772.5 NP_032798.1 1 1378
GenBank mRNA AK030759.1 - 1 1367
AK033660.1 - 1 1364
AK036611.1 - 1 1369
BC138634.1 - 1 1239 8861654
BC138635.1 - 1 1239 8861655
U22829.1 - 1 752

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
adenosine receptor signaling pathway
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
glial cell migration
response to mechanical stimulus
regulation of receptor activity
negative regulation of norepinephrine secretion
sensory perception of pain
signal transduction involved in regulation of gene expression
platelet activation
positive regulation of inositol trisphosphate biosynthetic process
G-protein coupled purinergic nucleotide receptor signaling pathway
regulation of vasodilation
eating behavior
positive regulation of ion transport
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hormone secretion
negative regulation of binding
positive regulation of penile erection
positive regulation of ERK1 and ERK2 cascade
cellular response to organic cyclic compound
protein localization to plasma membrane
relaxation of muscle
signal transducer activity
G-protein coupled receptor activity
ATP binding
A1 adenosine receptor binding
ADP binding
G-protein coupled purinergic nucleotide receptor activity
ATP-activated adenosine receptor activity
ADP-activated adenosine receptor activity
protein heterodimerization activity
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0