This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OTC SIRNA Assays ›
Gene Symbol:
Otc
Gene Name:
ornithine transcarbamylase
Gene Aliases:
AI265390, Sf, spf
Chromosome Location:
Chr.X: 10252297 - 10321024 on Build GRCm38
UniGene ID:
Mm.2611
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s71156

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Otc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008769.3 NP_032795.1 5 504
GenBank mRNA AK050930.1 - 5 505
BC024893.1 - 5 496 4196455
M17030.1 - 5 430

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_013078, K00001, BC088158, X01976, M11266, and others.

Gene Ontology Categories:

Function(s) Process(es)

urea cycle
cellular amino acid metabolic process
arginine biosynthetic process
ornithine metabolic process
ornithine catabolic process
cellular amino acid biosynthetic process
citrulline biosynthetic process
arginine biosynthetic process via ornithine
anion homeostasis
protein homotrimerization
ammonia homeostasis
ornithine carbamoyltransferase activity
phospholipid binding
amino acid binding
transferase activity
carboxyl- or carbamoyltransferase activity
phosphate ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0