This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OC90 SIRNA Assays ›
Gene Symbol:
Oc90
Gene Name:
otoconin 90
Gene Aliases:
Ocn-95, PLA2L, Pla2ll, oc95
Chromosome Location:
Chr.15: 65876050 - 65912411 on Build GRCm38
UniGene ID:
Mm.41776
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s70917

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Oc90
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010953.2 NP_035083.1 13 1093
GenBank mRNA AF091846.1 - 10 902
AF093591.1 - 13 1203
AK012981.1 - 13 1095
BC048716.1 - 11 964 6334417

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
lipid catabolic process
phospholipase A2 activity
structural molecule activity
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0