This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NTN1 SIRNA Assays ›
Gene Symbol:
Ntn1
Gene Name:
netrin 1
Gene Aliases:
AI561871, BC019633, Netrin-1
Chromosome Location:
Chr.11: 68209364 - 68386826 on Build GRCm38
UniGene ID:
Mm.39095
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s70847

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ntn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008744.2 NP_032770.2 7 1839
GenBank mRNA AF128865.1 - 6 1574
BC141294.1 - 6 1608 9055906
U65418.1 - 6 1555

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053731, XM_006246567, XM_006246568, AY028417.

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
apoptotic process
axonogenesis
axon guidance
positive regulation of cell proliferation
single organismal cell-cell adhesion
regulation of cell migration
negative regulation of axon extension
mammary gland development
anterior/posterior axon guidance
establishment of nucleus localization
inner ear morphogenesis
positive regulation of axon extension
mammary gland duct morphogenesis
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0