This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PROL1 SIRNA Assays ›
Gene Symbol:
Prol1
Gene Name:
proline rich, lacrimal 1
Gene Aliases:
Muc10
Chromosome Location:
Chr.5: 88317312 - 88328817 on Build GRCm38
UniGene ID:
Mm.200411
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s70171

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prol1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008644.2 NP_032670.2 3 431
GenBank mRNA AF247816.1 - 3 433
BC021158.1 - 3 435
BC042902.1 - 3 429 4924369
U37531.1 - 3, 4 431

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0