This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM89B SIRNA Assays ›
Gene Symbol:
Fam89b
Gene Name:
family with sequence similarity 89, member B
Gene Aliases:
1110021A21Rik, MMTV, Mtvr2
Chromosome Location:
Chr.19: 5728087 - 5729666 on Build GRCm38
UniGene ID:
Mm.391142
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s70158

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam89b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023166.2 NP_075655.2 1 396
NM_181452.2 NP_852117.2 1 396
GenBank mRNA AB012726.1 - 1 381
AF212920.1 - 1 378
AF212921.1 - 1 363
AK003883.1 - 1 385
BC013894.1 - 1 374 4217979
BC030163.1 - 1 367 3674980
BC056166.1 - 1 381 5715229
BM942073.1 - 1 322
CJ132526.1 - 1 396

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001015013, BC093377.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of SMAD protein import into nucleus
transcription corepressor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0