This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMP8 SIRNA Assays ›
Gene Symbol:
Mmp8
Gene Name:
matrix metallopeptidase 8
Gene Aliases:
BB138268
Chromosome Location:
Chr.9: 7558429 - 7568486 on Build GRCm38
UniGene ID:
Mm.16415
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s69942

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mmp8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008611.4 NP_032637.3 4 717
GenBank mRNA AK089234.1 - 4 688
AK089336.1 - 4 692
AK089381.1 - 4 689
AK089516.1 - 4 678
AK137468.1 - 4 689
AK154937.1 - 4 688
AK155740.1 - 4 688
AK155832.1 - 4 688
AK155901.1 - 4 691
AK170177.1 - 4 688
AK170388.1 - 4 690
AK170760.1 - 4 688
AK171168.1 - 4 688
AK171613.1 - 4 690
BC042742.1 - 4 698 5032770
U96696.1 - 4 683
Y13342.1 - 4 615

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
collagen catabolic process
metalloendopeptidase activity
serine-type endopeptidase activity
calcium ion binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0