This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLK1B21 SIRNA Assays ›
Gene Symbol:
Klk1b21
Gene Name:
kallikrein 1-related peptidase b21
Gene Aliases:
Klk21, mGk-21
Chromosome Location:
Chr.7: 44102292 - 44106583 on Build GRCm38
UniGene ID:
Mm.443292
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68850

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klk1b21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010642.2 NP_034772.1 4 639
GenBank mRNA AB039276.1 - 4 642
AK144221.1 - 4 606
BC012243.1 - 4 573 4167772
BC145383.1 - 4 595 9053941

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0