This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KHK SIRNA Assays ›
Gene Symbol:
Khk
Gene Name:
ketohexokinase
Gene Aliases:
-
Chromosome Location:
Chr.5: 30921436 - 30931248 on Build GRCm38
UniGene ID:
Mm.22451
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68725

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Khk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008439.3 NP_032465.2 3 349
GenBank mRNA BC013464.1 - 3 349 4235560
Y09335.1 - 3 328

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose metabolic process
response to sucrose
response to glucose
response to zinc ion
phosphorylation
response to insulin
carbohydrate phosphorylation
regulation of glycogen metabolic process
nucleotide binding
ketohexokinase activity
ATP binding
fructokinase activity
kinase activity
transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0