This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOXD3 SIRNA Assays ›
Gene Symbol:
HOXD3
Gene Name:
homeobox D3
Gene Aliases:
HOX1D, HOX4, HOX4A, Hox-4.1
Chromosome Location:
Chr.2: 176136921 - 176173102 on Build GRCh38
UniGene ID:
Hs.93574
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s6847

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HOXD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006898.4 NP_008829.3 3 871
XM_005246509.3 XP_005246566.1 4 1138
XM_005246510.4 XP_005246567.1 4 954
XM_005246511.3 XP_005246568.1 4 1562
XM_005246513.4 XP_005246570.1 4 1165
XM_006712477.2 XP_006712540.1 3 848
XM_011511065.2 XP_011509367.1 4 2942
XM_011511066.2 XP_011509368.1 4 1016
GenBank mRNA BC005124.2 - 3 872 3936607
BC008789.2 - 2 803 3628279

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell-matrix adhesion
Notch signaling pathway
anterior/posterior pattern specification
positive regulation of gene expression
glossopharyngeal nerve morphogenesis
thyroid gland development
positive regulation of neuron differentiation
embryonic skeletal system morphogenesis
cartilage development
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0