This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other JCHAIN SIRNA Assays ›
Gene Symbol:
Jchain
Gene Name:
immunoglobulin joining chain
Gene Aliases:
9530090F24Rik, AI323815, Igj, Jch
Chromosome Location:
Chr.5: 88519809 - 88527897 on Build GRCm38
UniGene ID:
Mm.1192
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68159

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Jchain
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152839.3 NP_690052.2 3 365
GenBank mRNA AB644392.1 - 3 247
AK078865.1 - 3 251
AK160344.1 - 3 317
BC006026.1 - 3 269 3491646
J00544.1 - 3 252
M90766.1 - 3 268

Back To Top

More Information


Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
glomerular filtration
humoral immune response
antibacterial humoral response
positive regulation of protein oligomerization
innate immune response
positive regulation of respiratory burst
single-stranded DNA binding
antigen binding
IgA binding
phosphatidylcholine binding
immunoglobulin receptor binding
protein homodimerization activity
peptidoglycan binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0