This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNRNPL SIRNA Assays ›
Gene Symbol:
HNRNPL
Gene Name:
heterogeneous nuclear ribonucleoprotein L
Gene Aliases:
HNRPL, P/OKcl.14, hnRNP-L
Chromosome Location:
Chr.19: 38836378 - 38852339 on Build GRCh38
UniGene ID:
Hs.644906
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s6742

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HNRNPL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005335.1 NP_001005335.1 3, 4 408
NM_001533.2 NP_001524.2 3, 4 642
XM_011526886.1 XP_011525188.1 3, 4 431
XM_011526887.1 XP_011525189.1 3, 4 475
XM_011526889.1 XP_011525191.1 3, 4 526
GenBank mRNA AB044547.1 - 3, 4 637
AK292115.1 - 3, 4 407
AK298384.1 - 3, 4 478
AK301062.1 - 3, 4 416
AK313652.1 - 3, 4 577
BC069184.1 - 3, 4 581
BI256815.1 - 3, 4 408
X16135.1 - 3, 4 566

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_032619, NM_001134760, XM_006228692, XM_006228691, BC169048.

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA splicing, via spliceosome
RNA processing
gene expression
nucleotide binding
RNA binding
protein binding
transcription regulatory region DNA binding
poly(A) RNA binding
pre-mRNA intronic binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0