This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR162 SIRNA Assays ›
Gene Symbol:
Gpr162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, BC031437, Grca
Chromosome Location:
Chr.6: 124858444 - 124864219 on Build GRCm38
UniGene ID:
Mm.2514
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s67002

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpr162
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013533.3 NP_038561.1 4 1882
GenBank mRNA AK134941.1 - 4 1696
AK144354.1 - 3 1651
AK144543.1 - 4 1669
AK158847.1 - 3 2774
AK171145.1 - 5 2066
BC016413.1 - 3 1312
BC031437.1 - 4 1882 4527410

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header