This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GAS1 SIRNA Assays ›
Gene Symbol:
Gas1
Gene Name:
growth arrest specific 1
Gene Aliases:
AW554192, Gas-1
Chromosome Location:
Chr.13: 60174405 - 60177535 on Build GRCm38
UniGene ID:
Mm.22701
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s66464

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gas1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008086.2 NP_032112.1 1 2410
GenBank mRNA BC055747.1 - 2 1505
BC058628.1 - 2 2050 6401957
X65128.1 - 1 2240

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of mesenchymal cell proliferation
cell cycle
cell cycle arrest
axon guidance
positive regulation of cell proliferation
regulation of smoothened signaling pathway
dorsal/ventral pattern formation
negative regulation of protein processing
programmed cell death
cerebellum morphogenesis
dorsal/ventral neural tube patterning
negative regulation of cell growth
outer ear morphogenesis
middle ear morphogenesis
odontogenesis
embryonic digit morphogenesis
camera-type eye development
negative regulation of apoptotic process
cell fate commitment
negative regulation of smoothened signaling pathway
positive regulation of smoothened signaling pathway
negative regulation of mitotic cell cycle
developmental growth
eye morphogenesis
embryonic morphogenesis
embryonic cranial skeleton morphogenesis
embryonic skeletal system development
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
palate development
regulation of ER to Golgi vesicle-mediated transport
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0