This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EPHB4 SIRNA Assays ›
Gene Symbol:
Ephb4
Gene Name:
Eph receptor B4
Gene Aliases:
AI042935, Htk, MDK2, Myk1, Tyro11
Chromosome Location:
Chr.5: 137350109 - 137374522 on Build GRCm38
UniGene ID:
Mm.34533
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s65582

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ephb4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159571.1 NP_001153043.1 7 1919
NM_010144.6 NP_034274.4 7 1892
GenBank mRNA AK046884.1 - 7 1857
AK049340.1 - 7 1893
AK132392.1 - 7 1892
AK143422.1 - 1862
BC013342.1 - 7 1580
BC016505.1 - 7 1844 3709926
BC090839.1 - 7 1669
U06834.1 - 8 1505

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003751156, XM_003751157.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell migration involved in sprouting angiogenesis
heart morphogenesis
protein phosphorylation
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
phosphorylation
peptidyl-tyrosine phosphorylation
protein autophosphorylation
ephrin receptor signaling pathway
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0